Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glyr1
Entrez Gene ID
74022
Gene Name
glyoxylate reductase 1 homolog (Arabidopsis)
Gene Aliases
2810419J22Rik, 3930401K13Rik, AW545332, Npac
Chromosome Location
Chr.16: 5013902 - 5049910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079814.1 NP_001073282.1 6 - 7 689 102
NM_028720.2 NP_082996.2 6 - 7 689 102
GenBank mRNA AK014456.1 - 6 - 7 629 102
AK040841.1 - 6 - 7 639 102
AK145277.1 - 6 - 7 640 102
AK150349.1 - 6 - 7 633 102
AK151832.1 - 6 - 7 636 102
AK152586.1 - 6 - 7 636 102
AK152887.1 - 6 - 7 633 102
AK159476.1 - 6 - 7 643 102
AK183942.1 - 3 - 4 274 102
AK188170.1 - 1 - 2 75 102
AK189966.1 - 2 - 3 122 102
AK204179.1 - 3 - 4 274 102
BC006893.1 - 6 - 7 623 102

View Full Product Details