The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079814.1 | NP_001073282.1 | 13 - 14 | 1347 | 76 |
NM_028720.2 | NP_082996.2 | 13 - 14 | 1329 | 76 | |
GenBank mRNA | AK014456.1 | - | 13 - 14 | 1269 | 76 |
AK040841.1 | - | 13 - 14 | 1279 | 76 | |
AK145277.1 | - | 13 - 14 | 1280 | 76 | |
AK150349.1 | - | 13 - 14 | 1273 | 76 | |
AK151832.1 | - | 13 - 14 | 1295 | 76 | |
AK152586.1 | - | 13 - 14 | 1295 | 76 | |
AK152887.1 | - | 13 - 14 | 1273 | 76 | |
AK159476.1 | - | 13 - 14 | 1283 | 76 | |
AK190428.1 | - | 1 - 2 | 151 | 76 | |
AK199843.1 | - | 1 - 2 | 72 | 76 | |
AK199856.1 | - | 1 - 2 | 73 | 76 | |
AK200495.1 | - | 1 - 2 | 72 | 76 | |
BC006893.1 | - | 13 - 14 | 1263 | 76 |