Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nphp3
Entrez Gene ID
74025
Gene Name
nephronophthisis 3 (adolescent)
Gene Aliases
3632410F03Rik, AI550417, C230078J01, D330020E01Rik, pcy
Chromosome Location
Chr.9: 104002544 - 104043811 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028721.3 NP_082997.3 16 - 17 2536 79
GenBank mRNA AK014393.1 - 1 - 2 96 79
AK133217.1 - 16 - 17 2537 79
AK173327.1 - 16 - 17 2115 79
AY259499.1 - 16 - 17 2473 79
BC115724.1 - 16 - 17 2063 79

View Full Product Details