The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028772.3 | NP_083048.1 | 6 - 7 | 985 | 89 |
GenBank mRNA | AK004755.1 | - | 6 - 7 | 986 | 89 |
BC024126.1 | - | 2 - 3 | 268 | 89 | |
BC089599.1 | - | 6 - 7 | 988 | 89 |