The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028775.3 | NP_083051.1 | 3 - 4 | 522 | 60 |
GenBank mRNA | AK004699.2 | - | 3 - 4 | 522 | 60 |
AK054324.1 | - | 3 - 4 | 346 | 60 | |
AK087069.1 | - | 3 - 4 | 510 | 60 | |
AK162947.1 | - | 3 - 4 | 906 | 60 | |
BC064733.1 | - | 3 - 4 | 539 | 60 |