Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scyl3
Entrez Gene ID
240880
Gene Name
SCY1-like 3 (S. cerevisiae)
Gene Aliases
1200016D23Rik, 6030457O16, AW214499, Pace1
Chromosome Location
Chr.1: 163927958 - 163955126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286002.1 NP_001272931.1 4 - 5 747 72
NM_001286003.1 NP_001272932.1 4 - 5 873 72
NM_028776.5 NP_083052.1 4 - 5 934 72
GenBank mRNA AK004809.2 - 4 - 5 674 72
AK044100.1 - 4 - 5 834 72
AK162937.1 - 4 - 5 748 72
BC043085.1 - 4 - 5 635 72
BC066800.1 - 4 - 5 619 72

View Full Product Details