Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scyl3
Entrez Gene ID
240880
Gene Name
SCY1-like 3 (S. cerevisiae)
Gene Aliases
1200016D23Rik, 6030457O16, AW214499, Pace1
Chromosome Location
Chr.1: 163927958 - 163955126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286002.1 NP_001272931.1 13 - 14 2397 71
NM_001286003.1 NP_001272932.1 13 - 14 2648 71
NM_028776.5 NP_083052.1 13 - 14 2623 71
GenBank mRNA AK004809.2 - 13 - 14 2363 71
AK030824.1 - 13 - 14 2598 71
AK044100.1 - 13 - 14 2609 71
AK047267.1 - 1 - 2 1404 71
AK050999.1 - 1 - 2 158 71
AK162937.1 - 13 - 14 2398 71
BC043085.1 - 13 - 14 2324 71
BC066800.1 - 13 - 14 2308 71

View Full Product Details