Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep72
Entrez Gene ID
74470
Gene Name
centrosomal protein 72
Gene Aliases
2610029E11Rik, 4933440J22Rik
Chromosome Location
Chr.13: 74036495 - 74062314 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028959.3 NP_083235.3 10 - 11 1688 66
GenBank mRNA AK011607.1 - 5 - 6 612 66
AK017134.1 - 10 - 11 1680 66
AK034892.1 - 10 - 11 1681 66
AK161033.1 - 6 - 7 1013 66
AK173189.1 - 3 - 4 3672 66
BC060686.1 - 9 - 10 2676 66
BC100549.1 - 9 - 10 1538 66

View Full Product Details