The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029103.3 | NP_083379.2 | 2 - 3 | 257 | 68 |
GenBank mRNA | AB009291.1 | - | 2 - 3 | 231 | 68 |
AK014338.1 | - | 2 - 3 | 235 | 68 | |
AK131997.1 | - | 2 - 3 | 258 | 68 | |
AK165646.1 | - | 2 - 3 | 233 | 68 | |
AK167580.1 | - | 2 - 3 | 257 | 68 | |
AK176996.1 | - | 2 - 3 | 151 | 68 | |
AK186239.1 | - | 2 - 3 | 151 | 68 | |
BC038901.1 | - | 2 - 3 | 245 | 68 | |
BC048616.1 | - | 1 - 2 | 320 | 68 |