Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqcd
Entrez Gene ID
75732
Gene Name
IQ motif containing D
Gene Aliases
4933433C09Rik, AI662646
Chromosome Location
Chr.5: 120586060 - 120607243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029408.2 NP_083684.1 1 - 2 72 75
GenBank mRNA AK017032.1 - 1 - 2 72 75
AK076996.1 - 1 - 2 79 75
BC100441.1 - 1 - 2 61 75
BC125429.1 - 1 - 2 66 75
BC137644.1 - 1 - 2 66 75

View Full Product Details