The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029410.3 | NP_083686.1 | 6 - 7 | 843 | 99 |
GenBank mRNA | AK017362.1 | - | 6 - 7 | 844 | 99 |
BC039807.1 | - | - | 1364 | 99 | |
BQ031795.1 | - | 1 - 2 | 99 |