Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl24
Entrez Gene ID
75785
Gene Name
kelch-like 24
Gene Aliases
1110046J11Rik, 4930429H24Rik, 6530402O20Rik, C85082
Chromosome Location
Chr.16: 20097457 - 20129226 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029436.3 NP_083712.4 3 - 4 1163 73
GenBank mRNA AK015239.1 - 3 - 4 1149 73
AK044892.1 - 3 - 4 1135 73
AK046446.1 - 1 - 2 241 73
AK163616.1 - 1 - 2 301 73
BC021407.1 - 1 - 2 717 73

View Full Product Details