Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift43
Entrez Gene ID
76411
Gene Name
intraflagellar transport 43
Gene Aliases
1700019E19Rik, R75064
Chromosome Location
Chr.12: 86082533 - 86162459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029601.3 NP_083877.1 3 - 4 236 72
GenBank mRNA AK006115.1 - 3 - 4 234 72

View Full Product Details