The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029727.2 | NP_084003.2 | 7 - 8 | 878 | 91 |
GenBank mRNA | AK008613.1 | - | 5 - 6 | 531 | 91 |
AK008663.1 | - | 7 - 8 | 874 | 91 | |
BC109335.1 | - | 6 - 7 | 781 | 91 | |
BC109336.1 | - | 7 - 8 | 812 | 91 |