The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029847.4 | NP_084123.2 | 7 - 8 | 1474 | 72 |
GenBank mRNA | AK013194.2 | - | 7 - 8 | 1426 | 72 |
AK019515.1 | - | 7 - 8 | 1440 | 72 | |
AK031147.1 | - | 7 - 8 | 1462 | 72 | |
AK032812.1 | - | 7 - 8 | 1440 | 72 | |
AK035464.1 | - | 7 - 8 | 1395 | 72 | |
AK039765.1 | - | 7 - 8 | 1470 | 72 | |
AK046378.1 | - | 7 - 8 | 1475 | 72 | |
AK083188.1 | - | 7 - 8 | 1428 | 72 | |
AK084090.1 | - | 7 - 8 | 1460 | 72 | |
AK086667.1 | - | 7 - 8 | 1433 | 72 | |
AK144817.1 | - | 7 - 8 | 1442 | 72 | |
AK165970.1 | - | 7 - 8 | 1442 | 72 | |
BC046790.1 | - | 7 - 8 | 1448 | 72 | |
BC058351.1 | - | 7 - 8 | 1448 | 72 |