Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bcl9
Entrez Gene ID
77578
Gene Name
B cell CLL/lymphoma 9
Gene Aliases
2610202E01Rik, 8030475K17Rik, A330041G23Rik, Gm130
Chromosome Location
Chr.3: 97203658 - 97298083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029933.4 NP_084209.3 10 - 10 4653 57
GenBank mRNA AK020724.1 - 1 - 1 647 57
AK084676.1 - 3 - 3 2580 57
AK147659.1 - 10 - 10 4653 57
AY296061.1 - 7 - 7 4169 57
BC019641.1 - 2 - 2 1100 57
BC137850.1 - 7 - 7 4141 57

View Full Product Details