The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029948.2 | NP_084224.1 | 3 - 4 | 1013 | 113 |
GenBank mRNA | AK019795.1 | - | 3 - 4 | 1014 | 113 |
AK167683.1 | - | 3 - 4 | 831 | 113 |