Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmab
Entrez Gene ID
77697
Gene Name
methylmalonic aciduria (cobalamin deficiency) type B homolog (human)
Gene Aliases
9130222L19Rik, ATR
Chromosome Location
Chr.5: 114431034 - 114444073 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029956.3 NP_084232.1 3 - 4 307 74
GenBank mRNA AK020286.1 - 3 - 4 307 74
AK034288.1 - 3 - 4 272 74
AK088785.1 - 3 - 4 299 74
AK142286.1 - 3 - 4 299 74
AK148421.1 - 3 - 4 342 74
BC022159.1 - 3 - 4 261 74
BC057558.1 - 3 - 4 307 74

View Full Product Details