The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142728.1 | NP_001136200.1 | 10 - 11 | 1533 | 84 |
NM_001142729.1 | NP_001136201.1 | 10 - 11 | 1452 | 84 | |
NM_030070.3 | NP_084346.2 | 10 - 11 | 1533 | 84 | |
GenBank mRNA | AK015292.1 | - | 10 - 11 | 1450 | 84 |
AK019545.1 | - | 10 - 11 | 1466 | 84 | |
AK029802.1 | - | 10 - 11 | 1534 | 84 | |
AK031279.1 | - | 10 - 11 | 1451 | 84 |