Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dlst
- Entrez Gene ID
-
78920
- Gene Name
- dihydrolipoamide S-succinyltransferase (E2 component of 2-oxo-glutarate complex)
- Gene Aliases
-
1600017E01Rik, 4632413C10Rik, 4930529O08Rik, DLTS
- Chromosome Location
-
Chr.12: 85110824 - 85134091 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK019713.2 |
- |
- |
1504 |
61 |
View Full Product Details