The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190406.1 | NP_001177335.1 | 2 - 3 | 763 | 91 |
NM_001190408.1 | NP_001177337.1 | 2 - 3 | 763 | 91 | |
NM_030228.3 | NP_084504.2 | 3 - 4 | 1021 | 91 | |
NM_144560.3 | NP_653146.1 | 3 - 4 | 1021 | 91 | |
GenBank mRNA | AF508323.1 | - | 1 - 2 | 665 | 91 |
AF508324.1 | - | 1 - 2 | 665 | 91 | |
AK019661.1 | - | 3 - 4 | 996 | 91 | |
AK052229.1 | - | 3 - 4 | 1022 | 91 | |
AK082864.1 | - | 2 - 3 | 731 | 91 | |
AK218035.1 | - | 1 - 2 | 136 | 91 | |
BC031785.1 | - | 2 - 3 | 713 | 91 |