The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030229.4 | NP_084505.2 | 3 - 4 | 611 | 119 |
GenBank mRNA | AK019868.2 | - | 3 - 4 | 612 | 119 |
AK032967.1 | - | - | 1245 | 119 | |
AK152517.1 | - | 3 - 4 | 584 | 119 | |
BC010793.1 | - | 3 - 4 | 614 | 119 |