The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053245.2 | NP_444475.2 | 5 - 6 | 835 | 64 |
GenBank mRNA | AF296412.1 | - | 5 - 6 | 784 | 64 |