Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d19
Entrez Gene ID
67249
Gene Name
TBC1 domain family, member 19
Gene Aliases
2810453K03Rik
Chromosome Location
Chr.5: 53809581 - 53909221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144517.4 NP_653100.1 16 - 17 1257 96
GenBank mRNA AK012836.1 - 16 - 17 1208 96
AK076128.1 - 16 - 17 1205 96
AK083084.1 - 16 - 17 1228 96
BC020138.1 - 16 - 17 1216 96

View Full Product Details