The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144533.2 | NP_653116.1 | 4 - 5 | 811 | 82 |
GenBank mRNA | AK041897.1 | - | 3 - 4 | 586 | 82 |
BC005737.1 | - | 4 - 5 | 811 | 82 | |
BC092086.1 | - | 4 - 5 | 520 | 82 |