Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slamf7
Entrez Gene ID
75345
Gene Name
SLAM family member 7
Gene Aliases
19A, 19A24, 4930560D03Rik, CRACC, CS1
Chromosome Location
Chr.1: 171632403 - 171653037 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144539.5 NP_653122.2 4 - 5 1092 87
GenBank mRNA AB196816.1 - 3 - 4 639 87
AB196817.1 - 3 - 4 639 87
AB196818.1 - 3 - 4 639 87
AB196819.1 - 3 - 4 639 87
AB196837.1 - 3 - 4 639 87
AF467909.1 - 3 - 4 639 87
AF467910.1 - 3 - 4 639 87
AF467911.1 - 3 - 4 639 87
AK030135.1 - 4 - 5 953 87
AK030148.1 - 4 - 5 1093 87
AK040678.1 - 3 - 4 720 87
AK089525.1 - 3 - 4 720 87
AK172665.1 - 3 - 4 686 87
BC011154.1 - 3 - 4 651 87

View Full Product Details