The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164566.1 | NP_001158038.1 | 3 - 4 | 282 | 105 |
NM_144882.4 | NP_659131.2 | 5 - 6 | 468 | 105 | |
GenBank mRNA | AK031266.1 | - | 5 - 6 | 423 | 105 |
AK032169.1 | - | 5 - 6 | 403 | 105 | |
BC014764.1 | - | 3 - 4 | 282 | 105 | |
BC021879.1 | - | 5 - 6 | 404 | 105 | |
BY069430.1 | - | 3 - 4 | 238 | 105 | |
BY739337.1 | - | 3 - 4 | 245 | 105 | |
EF410080.1 | - | - | 508 | 105 |