The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145100.3 | NP_659568.2 | 2 - 3 | 511 | 78 |
GenBank mRNA | AB041649.1 | - | 2 - 3 | 78 | |
AK012406.1 | - | 2 - 3 | 199 | 78 | |
AK045592.1 | - | 2 - 3 | 512 | 78 | |
AK082941.1 | - | 2 - 3 | 191 | 78 | |
BC058599.1 | - | 2 - 3 | 444 | 78 |