Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc25
Entrez Gene ID
67179
Gene Name
coiled-coil domain containing 25
Gene Aliases
2610528H13Rik, NSrp70
Chromosome Location
Chr.14: 65837302 - 65866604 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145944.4 NP_666056.1 6 - 7 504 121
GenBank mRNA AK012172.1 - 6 - 7 452 121
AK016286.1 - 6 - 7 416 121
AK078345.1 - 6 - 7 505 121
AK089982.1 - 6 - 7 429 121
AK139180.1 - 5 - 6 2880 121
BC025545.1 - - 452 121
BC128015.1 - 6 - 7 396 121
BC148235.1 - 6 - 7 403 121

View Full Product Details