The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152809.2 | NP_690022.2 | 12 - 13 | 1952 | 103 |
GenBank mRNA | AK032675.1 | - | 6 - 7 | 507 | 103 |
AK034842.1 | - | 2 - 3 | 133 | 103 | |
BC033601.1 | - | 9 - 10 | 967 | 103 |