The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007620.2 | NP_031646.2 | 1 - 2 | 414 | 78 |
GenBank mRNA | AK146805.1 | - | 1 - 2 | 342 | 78 |
BC012714.1 | - | 1 - 2 | 342 | 78 | |
BC158026.1 | - | 1 - 2 | 392 | 78 | |
BC158029.1 | - | 1 - 2 | 392 | 78 | |
U31966.1 | - | 1 - 2 | 413 | 78 |