The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007693.1 | NP_031719.1 | 2 - 3 | 249 | 59 |
GenBank mRNA | AK144504.1 | - | 2 - 3 | 243 | 59 |
BC026554.1 | - | 2 - 3 | 233 | 59 | |
M64278.1 | - | 2 - 3 | 249 | 59 |