The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007693.1 | NP_031719.1 | 7 - 8 | 1462 | 90 |
GenBank mRNA | AK144504.1 | - | 7 - 8 | 1456 | 90 |
BC026554.1 | - | 7 - 8 | 1446 | 90 | |
M64278.1 | - | 7 - 8 | 1462 | 90 |