Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crmp1
Entrez Gene ID
12933
Gene Name
collapsin response mediator protein 1
Gene Aliases
CRMP-1, DRP-1, Dpysl1, ULIP-3, Ulip3
Chromosome Location
Chr.5: 37242058 - 37292163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136058.2 NP_001129530.1 12 - 13 1887 61
NM_007765.4 NP_031791.3 12 - 13 1752 61
GenBank mRNA AK159049.1 - 12 - 13 1696 61
BC031738.1 - 12 - 13 1681 61
BC065046.1 - 12 - 13 1865 61
U72875.1 - 12 - 13 1497 61
Y09080.1 - 12 - 13 1727 61

View Full Product Details