Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mecom
Entrez Gene ID
14013
Gene Name
MDS1 and EVI1 complex locus
Gene Aliases
D630039M04Rik, Evi-1, Evi1, Jbo, Mds, Mds1, Mds1-Evi1, Prdm3, Znfpr1b1
Chromosome Location
Chr.3: 29951296 - 30509783 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007963.2 NP_031989.2 11 - 12 2308 65
GenBank mRNA AJ001482.1 - 10 - 11 2463 65
AK033517.1 - 11 - 12 1742 65
BC076620.1 - 11 - 12 2260 65
BC139762.1 - 9 - 10 1426 65
JQ665270.1 - 9 - 10 2428 65

View Full Product Details