The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008024.2 | NP_032050.2 | 1 - 1 | 1018 | 142 |
GenBank mRNA | AB221616.1 | - | 1 - 1 | 741 | 142 |
AK050949.1 | - | 1 - 1 | 240 | 142 | |
BC137805.1 | - | 1 - 1 | 763 | 142 | |
BC137806.1 | - | 1 - 1 | 763 | 142 |