Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Flot2
Entrez Gene ID
14252
Gene Name
flotillin 2
Gene Aliases
AI573412, Esa, reggie-2
Chromosome Location
Chr.11: 78037868 - 78060434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040403.1 NP_001035493.1 6 - 7 699 69
NM_001284227.1 NP_001271156.1 7 - 8 783 69
NM_001284228.1 NP_001271157.1 7 - 8 646 69
NM_008028.2 NP_032054.1 7 - 8 790 69
GenBank mRNA AK030199.1 - 6 - 7 662 69
AK149530.1 - 7 - 8 721 69
AK170557.1 - 6 - 7 700 69
AK188583.1 - 2 - 3 69
BC070423.1 - 7 - 8 748 69
U07890.1 - 7 - 8 660 69

View Full Product Details