The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160016.1 | NP_001153488.1 | 10 - 11 | 1227 | 67 |
NM_001160017.1 | NP_001153489.1 | 9 - 10 | 1178 | 67 | |
NM_008142.4 | NP_032168.1 | 10 - 11 | 1227 | 67 | |
GenBank mRNA | AB246709.1 | - | 8 - 9 | 921 | 67 |
AB246710.1 | - | 8 - 9 | 921 | 67 | |
AK156101.1 | - | 9 - 10 | 1175 | 67 | |
AK163844.1 | - | 10 - 11 | 1216 | 67 | |
AK191980.1 | - | 1 - 2 | 72 | 67 | |
AK209108.1 | - | 2 - 3 | 264 | 67 | |
AK218112.1 | - | 1 - 2 | 202 | 67 | |
BC013058.1 | - | 10 - 11 | 1218 | 67 | |
U29055.1 | - | 8 - 9 | 957 | 67 |