The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008216.3 | NP_032242.3 | 2 - 3 | 1180 | 99 |
GenBank mRNA | AK079729.1 | - | 2 - 3 | 1173 | 99 |
BC080281.1 | - | 2 - 3 | 1135 | 99 | |
U52524.2 | - | 2 - 3 | 1134 | 99 | |
U69695.1 | - | 2 - 3 | 1154 | 99 |