The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009904.2 | NP_034034.2 | 1 - 2 | 207 | 95 |
GenBank mRNA | BC050767.1 | - | 1 - 2 | 207 | 95 |
D14117.1 | - | 1 - 2 | 220 | 95 | |
D86323.1 | - | 1 - 2 | 220 | 95 | |
U08373.1 | - | 1 - 2 | 220 | 95 |