Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbln1
Entrez Gene ID
14114
Gene Name
fibulin 1
Gene Aliases
-
Chromosome Location
Chr.15: 85205944 - 85286294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010180.2 NP_034310.2 12 - 13 1575 58
GenBank mRNA AK035388.1 - 12 - 13 1528 58
AK083573.1 - 12 - 13 1578 58
AK085989.1 - 12 - 13 1518 58
AK086451.1 - 12 - 13 1576 58
AK137511.1 - 12 - 13 1522 58
AK159646.1 - 12 - 13 1530 58
BC007140.1 - 12 - 13 1519 58
EU543224.1 - 12 - 13 1513 58
X70854.1 - 12 - 13 1572 58

View Full Product Details