Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo1
Entrez Gene ID
14261
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
Chromosome Location
Chr.1: 162829561 - 162866548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010231.2 NP_034361.1 1 - 2 310 94
GenBank mRNA BC011229.1 - 1 - 2 310 94
D16215.1 - 1 - 2 200 94
U87456.1 - 1 - 2 295 94

View Full Product Details