The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204973.1 | NP_001191902.1 | 2 - 3 | 1744 | 119 |
NM_010238.3 | NP_034368.2 | 2 - 3 | 1563 | 119 | |
NR_037970.1 | 1 - 2 | 1298 | 119 | ||
GenBank mRNA | AB010246.1 | - | 1 - 2 | 68 | 119 |
AF045462.1 | - | 1 - 2 | 709 | 119 | |
AF318183.1 | - | 1 - 2 | 726 | 119 | |
AK147918.1 | - | 2 - 3 | 3093 | 119 | |
AK158970.1 | - | 2 - 3 | 1564 | 119 | |
AK168423.1 | - | 1 - 2 | 730 | 119 | |
AK168525.1 | - | 2 - 3 | 1435 | 119 | |
AK220444.1 | - | 1 - 2 | 711 | 119 | |
BC046809.1 | - | 1 - 2 | 351 | 119 | |
BC085251.1 | - | 2 - 3 | 1494 | 119 | |
BC138655.1 | - | 1 - 2 | 337 | 119 | |
BC138656.1 | - | 1 - 2 | 337 | 119 |