The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010266.2 | NP_034396.1 | 10 - 11 | 1160 | 78 |
GenBank mRNA | AF174583.1 | - | 10 - 11 | 1086 | 78 |
AF369880.1 | - | 10 - 11 | 1045 | 78 | |
AF369881.1 | - | 10 - 11 | 1045 | 78 | |
AF369882.1 | - | 10 - 11 | 1045 | 78 | |
AF369883.1 | - | 10 - 11 | 1045 | 78 | |
AF369884.1 | - | 10 - 11 | 1045 | 78 | |
AK041056.1 | - | 10 - 11 | 1099 | 78 | |
AK079419.1 | - | 10 - 11 | 968 | 78 | |
AK079459.1 | - | 10 - 11 | 77 | ||
AK139617.1 | - | - | 929 | 78 | |
AK157014.1 | - | 10 - 11 | 1122 | 78 | |
AK173138.1 | - | 10 - 11 | 1089 | 78 | |
BC076583.1 | - | 10 - 11 | 1073 | 78 |