The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077696.1 | NP_001071164.1 | 21 - 22 | 3078 | 92 |
NM_001284248.1 | NP_001271177.1 | 23 - 24 | 3590 | 92 | |
NM_001284249.1 | NP_001271178.1 | 22 - 23 | 3187 | 92 | |
NM_001284250.1 | NP_001271179.1 | 21 - 22 | 3075 | 92 | |
NM_010412.3 | NP_034542.3 | 21 - 22 | 3075 | 92 | |
GenBank mRNA | AF006602.2 | - | 20 - 21 | 2788 | 92 |
AF207748.1 | - | 20 - 21 | 2707 | 92 | |
AK122325.2 | - | 16 - 17 | 2117 | 92 | |
AK139202.1 | - | 22 - 23 | 3161 | 92 | |
AK145972.1 | - | 21 - 22 | 3041 | 92 | |
AK146483.1 | - | 21 - 22 | 3014 | 92 | |
AK155335.1 | - | 21 - 22 | 3049 | 92 | |
AK156008.1 | - | 15 - 16 | 3912 | 92 | |
AK157159.1 | - | 15 - 16 | 3912 | 92 | |
AK161956.1 | - | 1 - 2 | 207 | 92 | |
BC023309.1 | - | 12 - 13 | 1628 | 92 | |
BC049926.1 | - | 21 - 22 | 3009 | 92 | |
BC058554.1 | - | 23 - 24 | 3542 | 92 | |
BC060609.1 | - | 20 - 21 | 2869 | 92 |