The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010436.2 | NP_034566.1 | 1 - 1 | 750 | 97 |
GenBank mRNA | AK088040.1 | - | 1 - 1 | 772 | 97 |
BC005468.1 | - | 1 - 1 | 744 | 97 | |
BC010336.1 | - | 1 - 1 | 750 | 97 | |
X58069.1 | - | 1 - 1 | 753 | 97 |