Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmtf1
Entrez Gene ID
23857
Gene Name
cyclin D binding myb-like transcription factor 1
Gene Aliases
Dimp, Dmp1
Chromosome Location
Chr.5: 9118868 - 9161776 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110327.1 NP_001103797.1 15 - 16 2118 69
NM_011806.3 NP_035936.3 16 - 17 2328 69
GenBank mRNA AK029147.1 - 16 - 17 2270 69
AK046201.1 - 15 - 16 2713 69
AK078015.1 - 15 - 16 2022 69
AK160595.1 - 15 - 16 2091 69
BC045141.1 - 15 - 16 2042 69
BC060109.1 - 16 - 17 2250 69
U70017.1 - 16 - 17 2283 69

View Full Product Details