Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc6
Entrez Gene ID
23888
Gene Name
glypican 6
Gene Aliases
-
Chromosome Location
Chr.14: 116921001 - 117979529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079844.2 NP_001073313.1 8 - 9 1958 68
NM_011821.3 NP_035951.1 7 - 8 1928 68
GenBank mRNA AF105268.1 - 8 - 9 1651 68
AK132531.1 - 7 - 8 1665 68
AK149312.1 - 8 - 9 1725 68
AK164668.1 - 7 - 8 1930 68
BC023448.1 - 8 - 9 1372 68

View Full Product Details