Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ksr1
Entrez Gene ID
16706
Gene Name
kinase suppressor of ras 1
Gene Aliases
AW492498, B-KSR1, D11Bhm183e, D11Bhm184e, Ksr, RSU2, hb
Chromosome Location
Chr.11: 79013440 - 79146554 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013571.2 NP_038599.1 1 - 2 300 66
GenBank mRNA AK036566.1 - 1 - 2 356 66
AK164833.1 - 1 - 2 420 66
U43585.1 - 1 - 2 307 66
X81634.1 - 1 - 2 82 66

View Full Product Details