Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gyg
Entrez Gene ID
27357
Gene Name
glycogenin
Gene Aliases
AU017667, Gyg1
Chromosome Location
Chr.3: 20122084 - 20155266 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013755.3 NP_038783.1 5 - 6 716 125
GenBank mRNA AF114031.1 - 5 - 6 651 125
AK159576.1 - 5 - 6 697 125
AK206737.1 - 2 - 3 171 125
AK219866.1 - 2 - 3 171 125
BC029903.1 - 5 - 6 677 125

View Full Product Details